3,5-Dichloro-2-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

032510-1G
101833-892EA 160 USD
101833-892
3,5-Dichloro-2-(trifluoromethyl)pyridine ≥97%
3,5-Dichloro-2-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂Cl₂F₃N
MDL Number: MFCD11226518
CAS Number: 7655-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR