1-(2,2-Difluoropropyl)-4-fluorobenzene ≥97%

Supplier: Matrix Scientific

032507-1G
101833-886EA 275.38 USD
101833-886
1-(2,2-Difluoropropyl)-4-fluorobenzene ≥97%
1-(2,2-Difluoropropyl)-4-fluorobenzene

Hazard: Irritant

Formula: C₉H₉F₃
MDL Number: MFCD11226515
CAS Number: 58325-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR