1-Chloro-2-(2,2,2-trifluoroethyl)benzene ≥97%

032505-1G
101833-882EA 181.7 USD
101833-882
1-Chloro-2-(2,2,2-trifluoroethyl)benzene ≥97%
1-Chloro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆ClF₃
MDL Number: MFCD11226513
CAS Number: 145914-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR