(1-Trifluoromethylpentyl)cyclopentane ≥95%

Supplier: Matrix Scientific

032498-1G
101833-866EA 243.08 USD
101833-866
(1-Trifluoromethylpentyl)cyclopentane ≥95%
(1-Trifluoromethylpentyl)cyclopentane

Hazard: Irritant

Formula: C₁₁H₁₉F₃
MDL Number: MFCD11226529
CAS Number: 1099598-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR