3-Methyl-2-(trifluoromethyl)benzofuran ≥97%

Supplier: Matrix Scientific

032497-1G
101833-864EA 340 USD
101833-864
3-Methyl-2-(trifluoromethyl)benzofuran ≥97%
3-Methyl-2-(trifluoromethyl)benzofuran

Hazard: Irritant

Formula: C₁₀H₇F₃O
MDL Number: MFCD11226528
CAS Number: 1099598-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR