2-Fluoro-1,3-bis-(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-1,3-bis(trifluoromethyl)benzene2,6-Bis(trifluoromethyl)fluorobenzene

032495-5G
101833-860EA 198.46 USD
101833-860
2-Fluoro-1,3-bis-(trifluoromethyl)benzene ≥97%
2-Fluoro-1,3-bis-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₃F₇
MW: 232.1 g/mol
MDL Number: MFCD04972866
CAS Number: 887268-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR