3-Chloro-1,4-difluoro-2-(2,2,2-trifluoroethyl)benzene ≥97%

Supplier: Matrix Scientific

032494-500MG
101833-858EA 315.38 USD
101833-858
3-Chloro-1,4-difluoro-2-(2,2,2-trifluoroethyl)benzene ≥97%
3-Chloro-1,4-difluoro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₄ClF₅
MW: 230.57 g/mol
MDL Number: MFCD11226525
CAS Number: 1099598-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR