4-Fluoro-1-methyl-2-(2,2,2-trifluoroethyl)benzene ≥97%

Supplier: Matrix Scientific

032492-1G
101833-854EA 243.08 USD
101833-854
4-Fluoro-1-methyl-2-(2,2,2-trifluoroethyl)benzene ≥97%
4-Fluoro-1-methyl-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₉H₈F₄
MDL Number: MFCD11226524
CAS Number: 1099598-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR