2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine Hydrochloride

032490-1G
101833-850EA 287.69 USD
101833-850
2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride ≥95%
2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride

Hazard: Irritant

Formula: C₈H₈ClF₄N
MW: 229.61 g/mol
MDL Number: MFCD10565658
CAS Number: 886368-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR