2-Bromo-N-(4-chlorobenzyl)acetamide ≥95%

Supplier: Matrix Scientific

032487-1G
101833-844EA 215.38 USD
101833-844
2-Bromo-N-(4-chlorobenzyl)acetamide ≥95%
2-Bromo-N-(4-chlorobenzyl)acetamide

Hazard: Irritant

Formula: C₉H₉BrClNO
MW: 262.54 g/mol
MDL Number: MFCD11226535
CAS Number: 934185-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR