2-[4-Fluoro-3-(trifluoromethyl)phenyl]propan-2-ol ≥95%

Supplier: Matrix Scientific

032483-1G
101833-836EA 138.46 USD
101833-836
2-[4-Fluoro-3-(trifluoromethyl)phenyl]propan-2-ol ≥95%
2-[4-Fluoro-3-(trifluoromethyl)phenyl]propan-2-ol

Hazard: Irritant

Formula: C₁₀H₁₀F₄O
MDL Number: MFCD11226509
CAS Number: 534-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR