2,4-Dichloro-1,3,5-triazine ≥97%

Supplier: Matrix Scientific

032479-5G
102551-150EA 129.23 USD
102551-150
2,4-Dichloro-1,3,5-triazine ≥97%
2,4-Dichloro-1,3,5-triazine

Hazard: Irritant/Store in Freezer

Formula: C₃HCl₂N₃
MW: 149.9661 g/mol
Melting Pt: 50…56 °C
MDL Number: MFCD04115347
CAS Number: 2831-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR