3-(3-Chlorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride ≥97%

Supplier: Matrix Scientific

032475-250MG
101833-814EA 606.15 USD
101833-814
3-(3-Chlorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride ≥97%
3-(3-Chlorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃·2ClH
MW: 306.619 g/mol
MDL Number: MFCD11052320
CAS Number: 953411-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR