6-Methyl-3-(1H-pyrazol-5-yl)pyridin-2-ol ≥97%

Supplier: Matrix Scientific

032469-250MG
101833-802EA 303.08 USD
101833-802
6-Methyl-3-(1H-pyrazol-5-yl)pyridin-2-ol ≥97%
6-Methyl-3-(1H-pyrazol-5-yl)pyridin-2-ol

Hazard: Irritant

Formula: C₉H₉N₃O
MDL Number: MFCD01936098
CAS Number: 400077-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR