3-(Hydrazinomethyl)-N,N-dimethylaniline dihydrochloride ≥90%

Supplier: Matrix Scientific

032468-250MG
101833-800EA 300 USD
101833-800
3-(Hydrazinomethyl)-N,N-dimethylaniline dihydrochloride ≥90%
3-(Hydrazinomethyl)-N,N-dimethylaniline dihydrochloride

Hazard: Irritant-Harmful

Formula: C₉H₁₇Cl₂N₃
MDL Number: MFCD11052317
CAS Number: 1177344-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR