tert-Butyl piperidine-3-carboxylate ≥97%

Supplier: Matrix Scientific

032461-250MG
101833-784EA 167.69 USD
101833-784
tert-Butyl piperidine-3-carboxylate ≥97%
tert-Butyl piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD11041017
CAS Number: 301180-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR