2-(Dimethylamino)propan-1-ol ≥97%

Supplier: Matrix Scientific

032457-1G
101833-774EA 166.15 USD
101833-774
2-(Dimethylamino)propan-1-ol ≥97%
2-(Dimethylamino)propan-1-ol

Hazard: Irritant

Formula: C₅H₁₃NO
MDL Number: MFCD00014425
CAS Number: 15521-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR