2-Methyl-4-(thiophen-2-yl)butan-2-ol ≥90%

Supplier: Matrix Scientific

032456-500MG
101833-772EA 401.54 USD
101833-772
2-Methyl-4-(thiophen-2-yl)butan-2-ol ≥90%
2-Methyl-4-(thiophen-2-yl)butan-2-ol

Hazard: Irritant

Formula: C₉H₁₄OS
MDL Number: MFCD11052313
CAS Number: 778646-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR