Methyl-2-amino-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate hydrochloride ≥97%

Supplier: Matrix Scientific

032454-250MG
101833-768EA 303.08 USD
101833-768
Methyl-2-amino-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate hydrochloride ≥97%
Methyl-2-amino-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₁ClN₂O₄S
MW: 266.7 g/mol
MDL Number: MFCD11052312
CAS Number: 6497-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR