2-(tert-Butyl)-5-methyl-1H-indole ≥97%

Supplier: Matrix Scientific

032446-500MG
101833-750EA 290.77 USD
101833-750
2-(tert-Butyl)-5-methyl-1H-indole ≥97%
2-(tert-Butyl)-5-methyl-1H-indole

Hazard: Irritant

Formula: C₁₃H₁₇N
MDL Number: MFCD01463568
CAS Number: 69622-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR