5-Hydroxy-2-methoxybenzaldehyde ≥97%

Supplier: Matrix Scientific

032441-250MG
101833-740EA 113.85 USD
101833-740
5-Hydroxy-2-methoxybenzaldehyde ≥97%
5-Hydroxy-2-methoxybenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₈H₈O₃
MDL Number: MFCD03820046
CAS Number: 35431-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR