4-Bromo-1H-pyrazole-5-carbaldehyde ≥90%

Supplier: Matrix Scientific

032437-1G
101833-728EA 207.69 USD
101833-728
4-Bromo-1H-pyrazole-5-carbaldehyde ≥90%
4-Bromo-1H-pyrazole-5-carbaldehyde

Hazard: Irritant

Formula: C₄H₃BrN₂O
MW: 174.99 g/mol
MDL Number: MFCD00831607
CAS Number: 287917-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR