2-Hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylic acid methyl ester hydrate ≥97%

Supplier: Matrix Scientific

032426-1G
101833-702EA 64.62 USD
101833-702
2-Hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylic acid methyl ester hydrate ≥97%
2-Hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylic acid methyl ester hydrate

Hazard: Irritant

Formula: C₇H₅F₃N₂O₃·H₂O
MW: 240.137 g/mol
MDL Number: MFCD11100556
CAS Number: 1185300-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR