2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

032421-1G
101833-688EA 213.85 USD
101833-688
2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine ≥95%
2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆HCl₂F₄N
MDL Number: MFCD11100554
CAS Number: 1099598-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR