1-Fluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032419-1G
101833-684EA 190.77 USD
101833-684
1-Fluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1-Fluoro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD11100172
CAS Number: 93524-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR