3-(2,2,2-Trifluoroethyl)pyridine ≥95%

Supplier: Matrix Scientific

032416-1G
101833-680EA 246.15 USD
101833-680
3-(2,2,2-Trifluoroethyl)pyridine ≥95%
3-(2,2,2-Trifluoroethyl)pyridine

Hazard: Irritant

Formula: C₇H₆F₃N
MDL Number: MFCD11100551
CAS Number: 1099598-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR