(2,2,2-Trifluoroethyl)cyclopropane ≥95%

Supplier: Matrix Scientific

032412-1G
101833-670EA 255.38 USD
101833-670
(2,2,2-Trifluoroethyl)cyclopropane ≥95%
(2,2,2-Trifluoroethyl)cyclopropane

Hazard: Irritant

Formula: C₅H₇F₃
MDL Number: MFCD11100548
CAS Number: 3096-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR