2-Bromo-1,1,1,3-tetrafluoro-3-methylbutane ≥95%

Supplier: Matrix Scientific

032410-1G
101833-666EA 476.92 USD
101833-666
2-Bromo-1,1,1,3-tetrafluoro-3-methylbutane ≥95%
2-Bromo-1,1,1,3-tetrafluoro-3-methylbutane

Hazard: Irritant

Formula: C₅H₇BrF₄
MDL Number: MFCD11100547
CAS Number: 1099598-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR