1-Fluoro-4-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

032408-1G
101833-662EA 320 USD
101833-662
1-Fluoro-4-(3,3,3-trifluoropropyl)benzene ≥95%
1-Fluoro-4-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₈F₄
MDL Number: MFCD11100545
CAS Number: 1099598-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR