1-Fluoro-4-(5,5,5-trifluoropentyl)benzene ≥95%

Supplier: Matrix Scientific

032406-1G
101833-658EA 347.69 USD
101833-658
1-Fluoro-4-(5,5,5-trifluoropentyl)benzene ≥95%
1-Fluoro-4-(5,5,5-trifluoropentyl)benzene

Hazard: Irritant

Formula: C₁₁H₁₂F₄
MDL Number: MFCD11100543
CAS Number: 1099598-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR