2,6-Bis-(trifluoromethyl)naphthalene ≥95%

Supplier: Matrix Scientific

032404-1G
101833-654EA 364.62 USD
101833-654
2,6-Bis-(trifluoromethyl)naphthalene ≥95%
2,6-Bis-(trifluoromethyl)naphthalene

Hazard: Irritant

Formula: C₁₂H₆F₆
MW: 264.17 g/mol
MDL Number: MFCD00525619
CAS Number: 50318-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR