MATR032400

032400-1G
101833-646EA 64.08 USD
101833-646
MATR032400
Purity: 95%
CAS: 1099597-98-4
MDL Number: MFCD11100539
Molecular Formula: C8H6BrF3
Molecular Weight: 239,04
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR