5-(Trifluoromethyl)isoquinoline ≥95%

Supplier: Matrix Scientific

032397-1G
101833-640EA 347.69 USD
101833-640
5-(Trifluoromethyl)isoquinoline ≥95%
5-(Trifluoromethyl)isoquinoline

Hazard: Irritant

Formula: C₁₀H₆F₃N
MDL Number: MFCD11100536
CAS Number: 120568-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR