2,5-Dibromo-4-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

032386-1G
101833-616EA 164.62 USD
101833-616
2,5-Dibromo-4-(trifluoromethyl)pyridine ≥95%
2,5-Dibromo-4-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂Br₂F₃N
MDL Number: MFCD10699123
CAS Number: 1099597-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR