1-(2,2,2-Trifluoroethyl)-2,4-bis(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

032381-1G
101833-608EA 315.38 USD
101833-608
1-(2,2,2-Trifluoroethyl)-2,4-bis(trifluoromethyl)benzene ≥95%
1-(2,2,2-Trifluoroethyl)-2,4-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₀H₅F₉
MDL Number: MFCD11100530
CAS Number: 1099597-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR