1-Bromo-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032377-1G
101833-600EA 73.85 USD
101833-600
1-Bromo-2-(2,2,2-trifluoroethyl)benzene ≥95%
1-Bromo-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆BrF₃
MDL Number: MFCD09996898
CAS Number: 1027513-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR