(1,2,2,2-Tetrafluoroethyl)benzene ≥95%

032374-1G
101833-592EA 620.35 USD
101833-592
(1,2,2,2-Tetrafluoroethyl)benzene ≥95%
(1,2,2,2-Tetrafluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆F₄
MDL Number: MFCD11100525
CAS Number: 56424-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR