2,3-Bis(trifluoromethyl)-1,8-naphthyridine ≥95%

Supplier: Matrix Scientific

032373-500MG
101833-590EA 347.69 USD
101833-590
2,3-Bis(trifluoromethyl)-1,8-naphthyridine ≥95%
2,3-Bis(trifluoromethyl)-1,8-naphthyridine

Hazard: Irritant

Formula: C₁₀H₄F₆N₂
MDL Number: MFCD11100524
CAS Number: 1099597-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR