(Pentafluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032371-1G
101833-586EA 75.38 USD
101833-586
(Pentafluoroethyl)benzene ≥95%
(Pentafluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₅F₅
MDL Number: MFCD00511231
CAS Number: 309-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR