1,2-Difluoro-3-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

032369-1G
101833-582EA 283.08 USD
101833-582
1,2-Difluoro-3-(2,2,2-trifluoroethyl)benzene ≥95%
1,2-Difluoro-3-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₅F₅
MDL Number: MFCD11100522
CAS Number: 1099597-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR