5-(Trifluoromethyl)pyridine-2-carboxaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)picolinaldehyde

032348-1G
101833-524EA 300 USD
101833-524
5-(Trifluoromethyl)pyridine-2-carboxaldehyde ≥95%
5-(Trifluoromethyl)pyridine-2-carboxaldehyde

Hazard: Irritant/Air Sensitive/Refrigerate

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Melting Pt: 31…33 °C
MDL Number: MFCD07375388
CAS Number: 31224-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR