Cyclobutane-1,1-dicarboxylic acid ethyl ester ≥95%
Synonyms:
1-(Ethoxycarbonyl)cyclobutanecarboxylic acid
032342-1G
101833-508EA
535.02
USD
101833-508
Cyclobutane-1,1-dicarboxylic acid ethyl ester ≥95%
Cyclobutane-1,1-dicarboxylic acid ethyl ester
Hazard: Irritant
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol |
MDL Number:
MFCD10699115 CAS Number: 54450-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...