3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole ≥95%

Supplier: Matrix Scientific

032341-500MG
101833-506EA 150.77 USD
101833-506
3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole ≥95%
3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole

Hazard: Irritant

Formula: C₇H₄ClN₃S
MDL Number: MFCD03066187
CAS Number: 131986-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR