1H-Pyrrolo[3,2-b]pyridine-5-carboxylic acid

Supplier: Matrix Scientific

032332-250MG
101833-488EA 367.69 USD
101833-488
1H-Pyrrolo[3,2-b]pyridine-5-carboxylic acid
1H-Pyrrolo[3,2-b]pyridine-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MDL Number: MFCD09965906
CAS Number: 872355-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR