1H-Pyrrolo[3,2-c]pyridine-4-carbonitrile

Supplier: Matrix Scientific

032328-250MG
101833-480EA 480 USD
101833-480
1H-Pyrrolo[3,2-c]pyridine-4-carbonitrile
1H-Pyrrolo[3,2-c]pyridine-4-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MDL Number: MFCD09965899
CAS Number: 1040682-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR