1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile

Supplier: Matrix Scientific

032327-250MG
101833-478EA 300 USD
101833-478
1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MDL Number: MFCD09909831
CAS Number: 146767-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR