4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-B]pyridine-5-boronic acid pinacol ester

Supplier: Matrix Scientific

032325-250MG
101833-474EA 364.62 USD
101833-474
4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-B]pyridine-5-boronic acid pinacol ester
4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-B]pyridine-5-boronic acid pinacol ester

Hazard: Irritant

Formula: C₂₂H₃₆BClN₂O₂Si
MW: 434.89 g/mol
MDL Number: MFCD09965891
CAS Number: 1045857-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR