5-Bromo-2-(methylamino)-3-nitropyridine

Supplier: Matrix Scientific

Synonyms: 5-Bromo-N-methyl-3-nitropyridin-2-amine

032313-500MG
101833-444EA 326.15 USD
101833-444
5-Bromo-2-(methylamino)-3-nitropyridine
5-Bromo-2-(methylamino)-3-nitropyridine

Hazard: Irritant

Formula: C₆H₆BrN₃O₂
MW: 232.03 g/mol
MDL Number: MFCD07375006
CAS Number: 70232-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR