N,N-Dimethyl-3-nitropyridin-2-amine

Supplier: Matrix Scientific

032312-500MG
101833-442EA 541.54 USD
101833-442
N,N-Dimethyl-3-nitropyridin-2-amine
N,N-Dimethyl-3-nitropyridin-2-amine

Hazard: Irritant

Formula: C₇H₉N₃O₂
MDL Number: MFCD00956024
CAS Number: 5028-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR