N-(2-Chloro-3-formylpyridin-4-yl)pivalamide

Supplier: Matrix Scientific

032308-500MG
101833-434EA 526.15 USD
101833-434
N-(2-Chloro-3-formylpyridin-4-yl)pivalamide
N-(2-Chloro-3-formylpyridin-4-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
MDL Number: MFCD09965905
CAS Number: 338452-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR